Learn More
BINDER™ Reinforced Inner Chamber
Reinforced inner chamber for climate chambers and cooling incubation chambers.
Marca: BINDER™ 8012-2510
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
- Reinforced inner chamber for bulky and heavy samples.
- Weight capacity ranged from 100 to 200 kg, depending on model.
Specifica
KB 1060 | |
Stainless Steel | |
Chamber Rack | |
200 kg |